Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid-d, for NMR, 98 atom % D

Catalog No. AC166200500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166200500 50 g Glass Bottle
AC166202500 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166200500 Supplier Thermo Scientific Chemicals Supplier No. 166200500
Add to Cart

CAS: 758-12-3 | C2H4O2 | 61.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 758-12-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 61.058
MDL Number MFCD00051050
InChI Key QTBSBXVTEAMEQO-DYCDLGHISA-N
Synonym acetic acid-d, acetic acid-d1, monodeuteroacetic acid, acetic 2h acid, acetic acid-od, o-2h acetic acid, mono-deuteroacetic acid, ch3cood
PubChem CID 2723902
IUPAC Name deuterio acetate
SMILES CC(=O)O

Specifications

Physical Form Liquid
Color Colorless
Quantity 50 g
Packaging Glass Bottle
Flash Point 40°C
Refractive Index 1.3715
Specific Gravity 1.05
Deuterium (D) 97.5% min.
Water 0.4% max. (Coulometric)
Density 1.0500g/mL
Melting Point 15.0°C to 16.0°C
Boiling Point 116.0°C to 117.0°C
Linear Formula CH3CO2D
Formula Weight 61.06
Grade NMR
Chemical Name or Material Acetic acid-d, 98 atom % D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-059-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.