Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid-d4 99.5atom%D, TCI America™
SDP

Catalog No. A06375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A06375G Supplier TCI America Supplier No. A06375G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.08
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
PubChem CID 2723903
IUPAC Name (²H₃)ethan(²H)oic acid
SMILES [2H]OC(=O)C([2H])([2H])[2H]

Specifications

Melting Point 16°C
Color Colorless
Boiling Point 116°C
UN Number 2789
Quantity 5 g
Formula Weight 64.08
Physical Form Liquid
Chemical Name or Material Acetic Acid-d4 99.5atom%D
EINECSNumber (2)-0688
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.