Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acetic Acid, Glacial
SCI_ED

Catalog No. S25118B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
ACS Grade
ACS Grade, Safe-Cote
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25118B 500 mL ACS Grade
S25118A 500 mL ACS Grade, Safe-Cote
S25118 2.5 L ACS Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S25118B
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Assay Percent Range >95% w/v
Linear Formula CH3CO2H
Formula Weight 60.05
Grade ACS
Density 1.048g/mL
Quantity 500 mL
Packaging ACS Grade
Physical Form Liquid
Chemical Name or Material Acetic Acid, Glacial
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 8 (3), II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.