Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, glacial, J.T. Baker™

Catalog No. 14650388 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
HDPE bottle
PVC Coated Glass Bottle
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14-650-388 500 mL HDPE bottle
14-650-389 500 mL Poly bottle
14-650-233 2.5 L PVC Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-650-388 Supplier Avantor Supplier No. V19305
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Formula Weight 60.05
Quantity 500 mL
Packaging HDPE bottle
Physical Form Liquid
Chemical Name or Material Acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.