Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid, Glacial, TraceGrade, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600234 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-234 6 x 500 mL Poly Bottle
18-600-233 500 mL Poly Bottle
18-600-235 2.5 L Poly Bottle
18-600-236 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-234 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1008500MLCS6
Add to Cart
Add to Cart

A1008, 64-19-7, C2H4O2

Spectrum™ Chemical Acetic Acid, Glacial, TraceGrade also known under the name ethanoic acid, it is an organic compound and a colorless liquid. It has a distinctive sour taste and pungent smell. It is the main component of Vinegar after water (8% by volume). Classified as a weak acid, when concentrated it is corrosive and can attack the skin. Spectrum’s TraceGrade products are suitable for use trace metal analysis and provides the lowest levels of metals, organics, and particulates available; giving you improved analytical performance and minimizing repeated runs.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 100%
UN Number UN2789
Percent Purity 99%
Grade High Purity
Quantity 6 x 500 mL
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.