Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90, Reagents

Catalog No. 01335966
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-335-966 4 L Glass Bottle
01-335-969 1 L Glass Bottle
01-335-967 4 L Poly Bottle
01-335-965 4 L Safety Coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-335-966 Supplier Reagents Holdings Llc Supplier No. CA1007004C
Add to Cart
Add to Cart

Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 540-84-1
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid/2,2,4-Trimethylpentane
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9065
Grade Reagent
Chemical Name or Material Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.