Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, Puriss., 99-100%, Solstice

Catalog No. 60045756 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-756 4 x 5 L
60-000-27 1 L
60-000-28 2.5 L
60-000-29 5 L
60-045-757 6 x 1 L
60-045-755 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-045-756 Supplier Solstice Supplier No. 272214X5L
Add to Cart
Add to Cart

Puriss., 99-100%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Linear Formula CH3CO2H
UN Number UN2789
Formula Weight 60.05
Percent Purity 99 to 100%
Grade puriss.
Density 1.049g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 16% (92°F), 4% (59°F)
Flash Point 40°C (104°F)
Ignition Point 800°F
Vapor Density 2.07 (vs air)
Chemical Name or Material Acetic Acid
Show More Show Less
Hazard Statement P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
Precautionary Statement H226-H314
DOTInformation 3(8) / PGII
EINECSNumber 200-580-7
RTECSNumber AF1225000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.