Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Solstice

Catalog No. 60046216 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
60 kg
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
Plastic Pail
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-216 4 x 2.5 L Plastic Bottle
60-000-07 1 L Glass Bottle
60-000-09 2.5 L Glass Bottle
60-000-11 5 L Plastic Bottle
60-046-218 6 x 1 L Plastic Bottle
60-046-217 4 x 2.5 L Glass Bottle
60-000-10 30 kg Plastic Pail
60-000-12 60 kg Plastic Pail
60-000-06 1 L Plastic Bottle
60-000-08 2.5 L Plastic Bottle
60-046-219 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 60-046-216 Supplier Solstice Supplier No. 332094X2.5L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.2°C (literature)
Boiling Point 118°C
CAS Max % 100.0000%
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Percent Purity >99.8%
Grade ACS Reagent/Reagent ISO/Reagent Ph. Eur.
Density 1.049 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Flash Point 40°C
Packaging Plastic Bottle
Chemical Name or Material Acetic acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Acetic acid, glacial
EINECSNumber 200-580-7
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.