Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic acid, sodium salt, 99%, for HPLC, anhydrous

Catalog No. AC446880500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446880500 50 g Plastic bottle
AC446882500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446880500 Supplier Thermo Scientific Chemicals Supplier No. 446880500
Add to Cart

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324.0°C
Flash Point >250°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CO2Na
Quantity 50 g
Merck Index 15,8709
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity 99%
Grade HPLC
Insoluble Matter 0.005% max
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less
EINECSNumber 204-823-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.