Learn More
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Colorless |
| Linear Formula | CH3CO2H |
| UN Number | 2789 |
| Beilstein | 506007 |
| Formula Weight | 60.05g/mol |
| Percent Purity | ≥99% |
| Grade | TraceSELECT™ Ultra |
| Density | 1.049g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 4% (59°F), 16% (92°F) |
| Show More |
Safety and Handling
| Hazard Statement | P280-P305 + P351 + P338-P310 |
| Precautionary Statement | H226-H314 |
| EINECSNumber | 200-580-7 |
| RTECSNumber | AF1225000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
