Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic anhydride, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99% (GC), Solstice

Catalog No. 6000197 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-001-97 500 mL
60-001-95 1 L
60-001-96 2.5 L
60-046-228 6 x 1 L
60-046-227 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-001-97 Supplier Solstice Supplier No. 33214500ML
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99% (GC)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00008705
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Density 1.08g/mL
Flash Point 49°C (120.2°F)
Packaging Plastic Bottle
Linear Formula (CH3CO)2O
Quantity 500 mL
UN Number UN1715
Beilstein 385737
Formula Weight 102.09g/mol
Percent Purity ≥99% (GC)
Chemical Name or Material Acetic anhydride
Hazard Statement P210-P260-P280-P304 + P340 + P310-P305 + P351 + P338-P370 + P378
RTECSNumber AK1925000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.