Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride ACS AR, Macron Fine Chemicals™

Catalog No. MK24204 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-2420-4 4 L Glass
MK-2420-500 500 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-2420-4 Supplier Avantor Supplier No. 242008
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
PubChem CID 7918
ChEBI CHEBI:36610
IUPAC Name acetyl acetate
SMILES CC(=O)OC(=O)C

Specifications

Packaging Glass
Quantity 4 L
Formula Weight 102.09
Grade ACS
Chemical Name or Material Acetic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.