Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, ACS Reagent, Reagents

Catalog No. 01335865
Change view
Click to view available options
Quantity:
25 mL
125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-865 25 mL
01-335-866 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-335-865 Supplier Reagents Holdings Llc Supplier No. CA37000025C
Add to Cart
Add to Cart

Acetic Anhydride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
Synonym Acetyl acetate, Ethanoic Anhydride
IUPAC Name acetyl acetate
SMILES CC(=O)OC(C)=O

Specifications

Color Colorless
Boiling Point 139°C
Packaging Glass Bottle
Quantity 25 mL
Specific Gravity 1.08
Grade ACS
Physical Form Liquid
Chemical Name or Material Acetic Anhydride, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.