Learn More
Acetic Anhydride, Reagent, ACS, 97%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Specifications
Specifications
| CAS Max % | 100% |
| Boiling Point | 139°C (282.2 °F) |
| Packaging | Amber Glass Bottle |
| Quantity | 6 x 500 mL |
| UN Number | UN1715 |
| Residue after Evaporation | 0.003% |
| Percent Purity | 97% |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | Acetic Anhydride |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.