Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic Anhydride, Reagent, ACS, 97%, Spectrum™ Chemical
SDP

Catalog No. 18600243 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-243 4 x 2.5 L Amber Glass Bottle
18-600-247 125 mL Amber Glass Bottle
18-600-239 500 mL Amber Glass Bottle
18-600-241 2.5 L Amber Glass Bottle
18-600-240 6 x 500 mL Amber Glass Bottle
18-600-242 4 L Amber Glass Bottle
18-600-244 4 x 4 L Amber Glass Bottle
18-600-245 20 L Amber Glass Bottle
18-600-246 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-600-243 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A10152.5LTGLCS4
Add to Cart
Add to Cart

A1015, 108-24-7, C4H6O3

Spectrum™ Chemical Acetic Anhydride, Reagent, ACS , or ethanoic anhydride as it is otherwise known is a colorless, strong smelling liquid chemical compound that is the simplest isolatable acid anhydride and commonly used as a reagent for organic synthesis. Mainly used for acetylations, its main use is for the conversion of cellulose to cellulose acetate and in the synthesis of aspirin. As an ACS grade Reagent, Spectrum Chemical manufactured acetic anhydride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 108-24-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
InChI Key WFDIJRYMOXRFFG-UHFFFAOYSA-N
IUPAC Name acetyl acetate
SMILES CC(=O)OC(C)=O

Specifications

CAS Max % 100%
Boiling Point 139°C (282.2 °F)
Packaging Amber Glass Bottle
Quantity 4 x 2.5 L
UN Number UN1715
Residue after Evaporation 0.003%
Percent Purity 97%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Acetic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.