Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetic-d3 acid-d, for NMR, 99.5 atom % D

Catalog No. AC166210050
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166210050 5 mL
AC166210250 25 mL
AC166210500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166210050 Supplier Thermo Scientific Chemicals Supplier No. 166210050
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.08
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
PubChem CID 2723903
IUPAC Name deuterio 2,2,2-trideuterioacetate
SMILES [2H]OC(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 5 mL
Refractive Index 1.3709
Specific Gravity 1.13
Flash Point 40°C
Deuterium (D) 99.4% min.
Density 1.1300g/mL
Water 0.1% max. (Coulometric)
Melting Point 15.0°C to 16.0°C
Boiling Point 115.5°C
Linear Formula CD3CO2D
Beilstein 02, III, 203
Formula Weight 64.08
Grade NMR
Chemical Name or Material Acetic-d3 acid-d, 99.5 atom%D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.