Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018348 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8348 10 mL
11-101-8787 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8348 Supplier MilliporeSigma Supelco Supplier No. 9087210MLF
Add to Cart
Add to Cart

Acetone is a highly volatile and a flammable liquid, which comes under the class of ketones having an aromatic odor.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Vapor Pressure 184 mmHg (20°C)
Vapor Density 2 (vs. air)
Density 0.791 g/mL (at 25°C)
Flash Point -17°C
Physical Form Neat
Linear Formula CH3 COCH3
MDL Number MFCD00008765
Melting Point -94°C
Boiling Point 56°C (760 mmHg)
Type Analytical Standard
Quantity 10 mL
Formula Weight 58.08
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.