Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%, Solstice

Catalog No. 6000223 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
4 x 4 L
204 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-23 204 L Metal Drum
60-002-24 4 x 4 L Glass Bottle
60-002-25 4 x 4 L Poly Bottle
60-002-22 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-002-23 Supplier Solstice Supplier No. LP010204
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100.0000%
Density 0.79 g/cm3
Quantity 204 L
Flash Point −20°C
Packaging Metal Drum
Water 0.5% max.
UN Number UN1090
Beilstein 635680
Boiling Point 56°C
Solubility Information Miscible in Water at 20°C
Formula Weight 58.08
Percent Purity >99.5%
Grade ACS
Chemical Name or Material Acetone, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.