Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ACS Reagent, Reagents

Catalog No. 01338193
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
Safety Coated Glass
Steel Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-193 4 L Glass Bottle
01-338-198 1 L Glass Bottle
01-338-196 20 L Steel Pail
01-338-195 20 L HDPE Winpak
01-338-194 4 L Poly Bottle
01-338-192 4 L Safety Coated Glass
01-338-191 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-338-193 Supplier Reagents Holdings Llc Supplier No. C11025004C
Add to Cart
Add to Cart

Acetone, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.788
Boiling Point 56°C
Grade ACS
Chemical Name or Material Acetone, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.