Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RSOA00104C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Metal Can
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOA00104C 4 L Amber Glass Bottle
RSOA0010500 500 mL Amber Glass Bottle
RSOA00101C 1 L Amber Glass Bottle
RSOA00104A 4 L Natural Poly Bottle
RSOA001020F 20 L Cubitainer
RSOA001020E 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. RSOA00104C Supplier Ricca Chemical Company Supplier No. RSOA00104C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0003 meq/g
Titratable Base max. 0.0006 meq/g
Water max. 0.5%
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
Boiling Point 56°C
Percent Purity 99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
ShelfLife 60 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.