Learn More
Acetone-Alcohol, 1 + 1 Decolorizer Solution, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Physical Form | Solution |
| Chemical Name or Material | Acetone-Alcohol |
| Name Note | 1 + 1 Decolorizer Solution |
| CAS | 67-64-1, 64-17-5, 67-56-1 |
| CAS Min % | 49.0, 48.63 |
| CAS Max % | 51.0, 50.61 |
| Packaging | Natural Poly Bottle |
| Color | Colorless |
| Quantity | 500 mL |
| Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.