Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone - D6, deuteration degree min. 99.9% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1000210025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1000210025 25 mL Glass Bottle
M1000210100 100 mL Glass Bottle
M1000210009 10 x 0.75 mL Glass Ampule
M1000210010 10 mL Septum Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1000210025 Supplier MilliporeSigma Supplier No. 1.00021.0025

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Physical Form Liquid
Color Colorless
Viscosity 0.32mPa.s (20°C/68°F)
Packaging Glass Bottle
Quantity 25 mL
Vapor Pressure 233 hPa at 20°C (68°F)
Odor Characteristic
Assay Percent Range 99.9% min.
Formula Weight 64.12
Chemical Name or Material Acetone - D6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.