Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, FCC, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18600260 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-260 6 x 500 mL Amber Glass Bottle
18-600-253 100 mL Amber Glass Bottle
18-600-259 500 mL Amber Glass Bottle
18-600-254 1 L Amber Glass Bottle
18-600-261 4 L Amber Glass Bottle
18-600-255 4 L Poly Bottle
18-600-256 6 x 1 L Amber Glass Bottle
18-600-262 4 x 4 L Amber Glass Bottle
18-600-257 4 x 4 L Poly Bottle
18-600-258 20 L Steel Can
18-600-263 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-600-260 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1041500MLGLCS6
Add to Cart
Add to Cart

A1041, 67-64-1, C3H6O

Spectrum™ Chemical Acetone, FCC also known as 2-propanone, is an organic compound and a colorless, flammable liquid. Due to its miscibility with water, it serves as an important solvent. The U.S. Food and Drug Administration (FDA) has determined acetone is safe for use as an indirect food additive in adhesives and food-contact coatings and is regarded as a Generally Recognized as Safe (GRAS) substance at certain concentrations. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100%
Acidity 0.002%
Alkalinity 10 mg/kg
Quantity 6 x 500 mL
Packaging Amber Glass Bottle
Refractive Index 1.358-1.360
Water 0.5%
UN Number UN1090
Percent Purity 99.5 to 100.5%
Grade FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.