Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Solstice

Catalog No. 60090080 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-080 200 L
60-090-081 56 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-080 Supplier Solstice Supplier No. CS011200
Add to Cart
Add to Cart
Request a Quote

For HPLC, GC, pesticide residue analysis and spectrophotometry, >99.9% (excluding water)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
PubChem CID 180
ChEBI CHEBI:15347

Specifications

Quantity 200 L
Flash Point -20 °C
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.