Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Lab Grade, Innovating Science™
SCI_ED

Catalog No. S000492 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 L
3.8 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000492 1 L
S000491 3.8 L
S000493 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger: Highly flammable liquid and vapor, causes serious eye irritation, may cause drowsiness or dizziness.
Catalog No. S000492 Supplier Aldon Corporation Supplier No. IS10019
Add to Cart
Add to Cart
Danger: Highly flammable liquid and vapor, causes serious eye irritation, may cause drowsiness or dizziness.

Chemical Identifiers

CAS 67-64-1
Molecular Weight (g/mol) 58.08

Specifications

Quantity 1 L
Packaging HDPE
Boiling Point 56°C
Percent Purity 1
Grade Laboratory
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.