Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, meets ASTM D 329, Reagents

Catalog No. 01338188
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-188 4 L
01-338-189 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-338-188 Supplier Reagents Holdings Llc Supplier No. C11025504A
Add to Cart
Add to Cart

Acetone, meets ASTM D 329, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.788
Certifications/Compliance ASTM
Boiling Point 56°C
Chemical Name or Material Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.