Learn More
Description
Spectrum™ Chemical Acetone, NF is a low boiling, volatile organic compound. It is considered one of the five common excipient solvents suitable for pharmaceutical preparations, including water, methanol, dehydrated alcohol and ether. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 20 L |
| Packaging | Steel Can |
| Water | 0.5% |
| UN Number | UN1090 |
| Percent Purity | 99% |
| Grade | NF |
| Chemical Name or Material | Acetone |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
