Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Purified Suitable for Histology, Reagents

Catalog No. 01337614
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Bottle
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-614 4 L Poly Bottle
01-337-620 10 L Steel Pail
01-337-619 1 L Glass Bottle
01-337-617 20 L HDPE Winpak
01-337-616 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-614 Supplier Reagents Holdings Llc Supplier No. C21025004A
Add to Cart
Add to Cart

Acetone, Purified Suitable for Histology, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.788
Boiling Point 56°C
Grade Pure
Chemical Name or Material Acetone, Purified Suitable for Histology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.