Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Solstice

Catalog No. 60045556 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-556 20 kg Stainless Steel Can
60-045-553 1 L Poly Bottle
60-045-555 2.5 L Glass Bottle
60-045-554 2.5 L Poly Bottle
60-045-562 6 x 1 L Poly Bottle
60-045-559 4 x 2.5 L Glass Bottle
60-045-558 4 x 2.5 L Poly Bottle
60-045-560 4 x 5 L Poly Bottle
60-045-552 155 kg Metal Drum
60-045-561 5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 60-045-556 Supplier Solstice Supplier No. 2420120KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C
Quantity 20 kg
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Stainless Steel Can
Physical Form Liquid
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Boiling Point 55°C to 57°C
Formula Weight 58.08g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Acetone, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
missing translation for 'healthHazard2' P210-P305 + P351 + P338-P370 + P378-P403 + P235
missing translation for 'healthHazard3' H225-H319-H336
DOTInformation Class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.