Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetone, Reagent, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18600283 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-283 4 L Amber Glass Bottle
18-600-279 100 mL Amber Glass Bottle
18-600-280 500 mL Amber Glass Bottle
18-600-282 1 L Amber Glass Bottle
18-600-281 6 x 500 mL Amber Glass Bottle
18-600-284 4 L Poly Bottle
18-600-285 6 x 1 L Amber Glass Bottle
18-600-286 4 x 4 L Amber Glass Bottle
18-600-287 4 x 4 L Poly Bottle
18-600-288 20 L Steel Can
18-600-289 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-600-283 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A10204LTGL
Add to Cart
Add to Cart

A1020, 67-64-1, C3H6O

Spectrum™ Chemical Acetone, Reagent, ACS is colorless, flammable, liquid. Due to it's micsibilaty with water, it serves as an important solvent. Used as a polar aprotic solvent in a wide range of organic reactions, acetone does not form an azeotrope with water. Thanks to it's low cost and volatility, it is also commonly used to clean laboratory glassware. Another characteristic of Acetone is that it can be cooled with dry ice to -78°C without freezing allowing this solvent to be used to conduct reactions at low temperature. As an ACS grade Reagent, Spectrum Chemical manufactured Acetone is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
Titratable Acid 0.0003 meq/g
Titratable Base 0.0006 meq/g
Water 0.5%
UN Number UN1090
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Acetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.