Learn More
Acetone, Reagent, ACS, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Acetone, Reagent, ACS is colorless, flammable, liquid. Due to it's micsibilaty with water, it serves as an important solvent. Used as a polar aprotic solvent in a wide range of organic reactions, acetone does not form an azeotrope with water. Thanks to it's low cost and volatility, it is also commonly used to clean laboratory glassware. Another characteristic of Acetone is that it can be cooled with dry ice to -78°C without freezing allowing this solvent to be used to conduct reactions at low temperature. As an ACS grade Reagent, Spectrum Chemical manufactured Acetone is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 4 x 4 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.001% |
| Titratable Acid | 0.0003 meq/g |
| Titratable Base | 0.0006 meq/g |
| Water | 0.5% |
| UN Number | UN1090 |
| Percent Purity | 99.5% |
| Grade | Reagent |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.