Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, HPLC Grade, Cenmed™
SDP

Catalog No. 21520122
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
21-520-122 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-122 Supplier Cen-Med Enterprises Supplier No. 393405HPLC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified, dried and distilled in glass. Acetonitrile, HPLC Grade, Cenmed™ is a high-purity solvent for use in analytical and preparative separations. It is suitable for high resolution gas chromatography, trace organic analysis, and critical and routine HPLC.

Acetonitrile for use in high performance liquid chromatography (HPLC). This solvent conforms to the specifications defined by the American Chemical Society (ACS). ACS Grade, HPLC Grade.

This ultra-pure product is packaged in four 4L amber glass bottles to protect the integrity of the solvent from UV light. Glass GC analysis of this product using an electron capture detector shows no peaks with a response greater than 10 ppt as Heptaehlor Epoxide.

  • HPLC mobile phase for analytical and preparative separations
  • High UV transmittance and low evaporation residue
  • Suitable for gradient and isocratic conditions
  • Glass purified and glass distilled

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
SMILES CC#N

Specifications

Melting Point 6°C
Quantity 4 x 4 L
Packaging Amber Glass Bottle
Physical Form Liquid
Linear Formula CH3CN
UN Number 1648
Boiling Point 81.6°C
Formula Weight 41.05g/mol
Grade Mobile phase for HPLC applications
Chemical Name or Material Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.