Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.08% TFA, LabReady™ Solvent Blend, for HPLC, Solstice

Catalog No. 60048260 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-260 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-260 Supplier Solstice Supplier No. 5734
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blends

  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Solstice Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Solstice LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Solstice's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1 , 75-05-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

CAS Min % 0.04, >99.00
CAS Max % 0.10
Density 0.777 g/cm3
Quantity 4 x 4 L
Flash Point The physical data is that of the main component..6°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 82°C
Grade HPLC
Hazard Category Danger
Hazard Statement H225+H302+H332+H319+H370
Precautionary Statement P280+P271+P270+P264+P260+P243+P242+P241+P240+P233+P210
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.