Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% acetic acid, CHROMASOLV™ LC-MS, LabReady™ Solvent Blend, Solstice

Catalog No. 60048231 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-231 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-231 Supplier Solstice Supplier No. 346784X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8 , 64-19-7
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 99.90, 0.10
Density 0.7770 g/cm3
Quantity 4 x 4 L
Flash Point 19°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 83°C
Grade LC-MS
Hazard Category Danger
Hazard Statement H370+H319+H302+H332+H225
Precautionary Statement P210+P233+P240+P241+P242+P243+P260+P264+P270+P271+P280
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.