Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% Acetic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Solstice

Catalog No. 6000048 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-000-48 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-000-48 Supplier Solstice Supplier No. LC4622.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-05-8 , 76-05-1
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Boiling Point 83°C
CAS Max % 99.9000%, 0.1000%
UN Number UN1648
Density 0.7770 g/cm3
Grade HPLC
Packaging Glass Bottle, Gl45
Quantity 4 x 2.5 L
Flash Point 19°C
Chemical Name or Material Acetonitrile with 0.1% Acetic Acid, LabReady™ Solvent Blend, for LC-MS and HPLC
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.