Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% formic acid, CHROMASOLV™ LC-MS, Solstice

Catalog No. 60048215 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
Packaging:
GL45 Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-215 2.5 L Glass Bottle
60-048-223 4 x 2.5 L GL45 Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-215 Supplier Solstice Supplier No. 346684X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ LC-MS, LabReady™ Solvent Blend

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

pH 4.1
Melting Point -46°C
Color Colorless
Density 0.7767 g/mL at 25°C
Quantity 2.5 L
Ignition Point 524°C
Odor Sweet ether-like
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 97mmHg
UN Number 1648
Boiling Point 82°C
Percent Purity 99.5 to 100%
Grade LC-MS
Chemical Name or Material Formic acid in acetonitrile
Show More Show Less
Hazard Category Danger
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.