Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile with 0.1% trifluoroacetic acid, Solstice

Catalog No. 60048232 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-232 4 x 4 L Glass Bottle
60-048-330 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-232 Supplier Solstice Supplier No. 349764X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ LC-MS, LabReady™ Solvent Blend

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Quantity 4 x 4 L
Flash Point 6°C (42.8°F)
Packaging Glass Bottle
UN Number UN1648
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile
Molecular Weight (g/mol) 41.05
Percent Purity >99.5% (GC)
Chemical Name or Material Acetonitrile with 0.1% trifluoroacetic acid
Hazard Category Danger
Hazard Statement P210-P305 + P351 + P338
EINECSNumber 200-835-2
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.