Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, GR, ACS, MilliporeSigma™

Catalog No. MAX01493 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Steel Barrel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01493 20 L Steel Barrel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAX01493 Supplier MilliporeSigma Supplier No. AX01493
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Color Colorless
Appearance Clear
Density 0.786g/cc at 20°C
Quantity 20 L
Flash Point 2°C
Ignition Point 524°C
Odor Ether-Like
Packaging Steel Barrel
Titratable Acid >8μeq/g
Vapor Pressure 97 hPa (20°C)
Viscosity 0.316 mPa.s at 25°C
UN Number UN 1648
Water >0.3%
Boiling Point 81.6°C (1013 hPa)
Formula Weight 41.05
Percent Purity <99.5%
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Danger
Hazard Statement H225: Highly flammable liquid and vapor. H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled. H319: Causes serious eye irritation.
Precautionary Statement P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P240 Ground/bond container and receiving equipment. P302 + P352 IF ON SKIN: Wash with plenty of soap and water. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P233 Store in a well-ventilated place. Keep container tightly closed
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.