Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
4x25mL
Packaging:
AcroSeal™ Glass Bottle
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide,cyanomethane,ethanenitrile,methanecarbonitrile,ethyl nitrile,methane, cyano,acetonitril,cyanure de methyl,methylkyanid,mecn
6342
CHEBI:38472
acetonitrile
CC#N
Acetic Acid 0.002 % max.
Assay Percent Range 99.9%
Linear Formula CH3CN
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Physical Form Liquid
Percent Purity 99.9%
Grade Extra Dry
Density 0.7810g/mL
Identification Pass Test
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3430 to 1.3450
Residue after Evaporation 0.0005 % max.
Solubility Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Specific Gravity 0.781
Water 0.001% Max. (Coulometric)
Flash Point 2°C
Viscosity 0.36 mPa.s (20°C)
Boiling Point 81.0°C to 82.0°C
Color Colorless
Melting Point -46.0°C
Quantity 4x25mL
Chemical Name or Material Acetonitrile
Show More Show Less