Learn More
Acetonitrile, For pesticide residue analysis, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| Color | Colorless |
| Density | 0.786g/mL at 25°C |
| Quantity | 4 x 4 L |
| Explosion Limit | 0.16 |
| Flash Point | 2°C (35.6°F) |
| Ignition Point | 973°F |
| Packaging | Glass Bottle |
| Vapor Pressure | 72.8 mmHg (20°C) |
| Linear Formula | CH3CN |
| UN Number | UN1648 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P305 + P351 + P338 |
| DOTInformation | 3 / PGII |
| RTECSNumber | 200-835-2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.