Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, ACS, 99.5+%

Catalog No. p-4408220
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36423AP 500 mL
AA36423K2 1 L
AA36423M1 4 x 1 L
AA36423K7 4 L
AA36423M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA36423AP Supplier Thermo Scientific Chemicals Supplier No. 036423AP
Only null left
Add to Cart

CAS: 75-05-8 | C2H3N | 41.053 g/mol

It is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. It is used as a mobile phase in HPLC and in LC-MS. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point °C to 48°C to°C to 45°C
Color Colorless
Appearance Colorless liquid
Density 0.786 g/cm3 at 20°C
Quantity 500 mL
Flash Point 5°C (41°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range >99.5%
UN Number 1648; Colorless
Beilstein 741857
Solubility Information Miscible with water and methanol.
Boiling Point 80°C to 82°C
Formula Weight 41.05
Grade ACS
Chemical Name or Material Acetonitrile, For analysis ACS
Show More Show Less
Hazard Category H225-H302+H312+H332-H319
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 200-835-2
RTECSNumber AL7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.