Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, ACS, MilliporeSigma™

Catalog No. MAX0149PP20 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
200 L
Packaging:
Glass Bottle
Metal Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX0149PP20 200 L Metal Drum
MAX01496 500 mL Glass Bottle
MAX014920 200 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MAX0149PP20 Supplier MilliporeSigma Supplier No. AX0149PP20
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Quantity 200 L
Packaging Metal Drum
Physical Form Liquid
Assay Percent Range 99.5% min.
Formula Weight 41.05
Chemical Name or Material Acetonitrile, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.