Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%, Honeywell Burdick & Jackson™

Catalog No. 6000227 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Septum-sealed Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-27 6 x 1 L Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-002-27 Supplier Honeywell Chemicals Supplier No. AS0171

Anhydrous, septum-sealed, for applications requiring low water content, >99.9%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Boiling Point 82°C
Quantity 6 x 1 L
Flash Point 6°C
Packaging Septum-sealed Bottle
Physical Form anhydrous
UN Number UN1648
Beilstein 741857
Formula Weight 41
Percent Purity >99.9%
Chemical Name or Material Acetonitrile, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.