Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, for applications requiring low water content, meets UV Specifications, >99.9%, Solstice

Catalog No. 6000278 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
200 L
Packaging:
Returnable Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-78 56 L Returnable Steel Drum
60-002-75 200 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-002-78 Supplier Solstice Supplier No. CS01756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Absorbance 0.05 AU max. at 200 nm, 0.04 AU max. at 210 nm, 0.01 AU max. at 225 nm, 0.005 AU max. at 250 nm, 0.005 AU max. at 350 nm
CAS Max % 100.0000%
Density 0.7822 g/cm3
Quantity 56 L
Flash Point 6°C
Packaging Returnable Steel Drum
UN Number UN1648
Water 10 ppm max.
Beilstein 741857
Boiling Point 82°C
Solubility Information Miscible in Water at 20°C
Formula Weight 41
Percent Purity >99.9%
Chemical Name or Material Acetonitrile, B&J Brand™, for applications requiring low water content, meets UV Specifications, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.