Learn More
Acetonitrile, for GC, for pesticide residue analysis, non-spectrophotometric grade, ≥99.9%, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.7822 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | 6°C |
| Packaging | Glass Bottle |
| Water | 0.05% max. |
| UN Number | UN1648 |
| Beilstein | 741857 |
| Boiling Point | 82°C |
| Solubility Information | Miscible in Water at 20°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.