Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, LC-MS, for LC-MS and HPLC, >99.9%, Solstice
BIOPHARMA

Catalog No. 6000263 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 L
4 x 2.5 L
6 x 1 L
Packaging:
GL45 Glass Bottle
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-002-63 6 x 1 L GL45 Glass Bottle
60-002-62 4 L Glass Bottle
60-002-61 4 x 2.5 L GL45 Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-002-63 Supplier Solstice Supplier No. LC0151
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Quantity 6 x 1 L
Flash Point 6°C
Packaging GL45 Glass Bottle
Water 0.01% max.
UN Number UN1648
Beilstein 741857
Solubility Information Miscible in Water at 20°C
Boiling Point 82°C
Formula Weight 41
Percent Purity >99.9%
Grade LC-MS
Chemical Name or Material Acetonitrile, B&J Brand™ LC-MS, for LC-MS and HPLC, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.