Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, BioSyn™, for biosynthesis and applications requiring low water content, meets UV specifications, >99.9%, Solstice

Catalog No. 60048327 Shop All Solstice Research Chemicals Products
Catalog No. 60-048-327 Supplier Solstice Supplier No. BB0174
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym Methyl Cyanide, Cyanomethane
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.00
Density 0.7822 g/cm3
Flash Point 6°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 82°C
Grade Low Water
Hazard Category Danger
Hazard Statement H370+H319+H302+H332+H225
Precautionary Statement P210+P233+P240+P241+P242+P243+P260+P264+P270+P271+P280
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.