Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile (Certified ACS), Fisher Chemical™ PROMO

Catalog No. A214S24 Shop All Fisherbrand Products
Change view
Click to view available options
Acetonitrile (ACS); Packaging: Glass Bottle; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A214S24 Acetonitrile (ACS); Packaging: Glass Bottle; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A214S24

Synonym: Cyanomethane, Methyl Cyanide, Ethanenitrile

  • Meet or exceeds the latest ACS specifications
  • Actual lot of analysis on label
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Decomposition Information >500°C
Density 0.781 g/cm3
Quantity 4 L
Flash Point 12.8°C (55°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.5%
Residue after Evaporation 0.005% Maximum
Titratable Acid 0.008 mEq/g
Linear Formula CH3CN
Titratable Base 0.0006 mEq/g
Vapor Pressure 97 mbar at 20°C
Viscosity 0.36 cP at 20°C
UN Number 1648
Water 0.3%
Boiling Point 81°C
Formula Weight 41.05
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Acetonitrile
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement Flammable Liquid and Vapor.
Precautionary Statement Causes skin irritation. Causes eye irritation. May cause respiratory irritation. May cause damage to organs.
DOTInformation DOT Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.