Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Solstice
BIOPHARMAGSA_VA

Catalog No. 6000247 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-002-47 4 x 4 L
60-002-42 1 L
60-002-43 2.5 L
60-002-46 6 x 1 L
60-002-40 100 mL
60-046-486 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 60-002-47 Supplier Solstice Supplier No. 348514X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, gradient grade, for HPLC,≥99.9%

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Absorbance HPLC-gradient/254nm ≤0.5mAU
Density 0.786g/mL at 25°C
Quantity 4 x 4 L
Refractive Index n20/D 1.344
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
Beilstein 741857
Boiling Point 81°C to 82°C
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Gradient
Chemical Name or Material Acetonitrile, for HPLC
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
EINECSNumber 200-835-2
RTECSNumber AL7700000
Recommended Storage RT
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.