Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetonitrile, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18608410 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-410 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-608-410 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP416200LTBL

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.05

Specifications

CAS Max % 1
Quantity 200 L
Packaging Drum
Residue after Evaporation 5 ppm
Titratable Acid 8 μeq/g
Titratable Base 0.6 μeq/g
Water 0.0002
Boiling Point 81.6°C
Percent Purity 0.999
Grade ACS/HPLC
Chemical Name or Material Acetonitrile
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.